Learn More
Biomatik Corporation Human Interleukin 12A (IL12A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54499
Encompass_Preferred
The Human Interleukin 12A (IL12A) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 11.9 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P29459. NCBI Gene ID: 3592. Restriction: For Research Use Only.
Catalog No. 50-150-1359