Learn More
Biomatik Corporation Mouse Interleukin 13 (IL13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54513
Encompass_Preferred
Mouse Interleukin 13 (IL13) ELISA Kit for detecting IL13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P20109. Gene ID: 16163. Target Synonyms: ALRH; BHR1; P600; IL-13; NC30. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1369