Learn More
Biomatik Corporation Rat Interleukin 13 (IL13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54515
Encompass_Preferred
Rat Interleukin 13 (IL13) ELISA Kit for detecting IL13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P42203. Gene ID: 116553. Target Synonyms: ALRH; BHR1; P600; IL-13; NC30. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1370