Learn More
Biomatik Corporation Rat Interleukin 17 (IL17) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54535
Encompass_Preferred
Rat Interleukin 17 (IL17) ELISA Kit for detecting IL17 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q61453. Gene ID: 301289. Target Synonyms: IL17A; CTLA8; IL-17A; Cytotoxic T-Lymphocyte-Associated Protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1381