Learn More
Biomatik Corporation Human Interleukin 18 Receptor Accessory Protein (IL18RAP) ELISA Kit, 96 tests
Catalog No. 501501394
Encompass_Preferred
The Human Interleukin 18 Receptor Accessory Protein (IL18RAP) ELISA Kit allows for quantitative determination in tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.051 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O95256. NCBI Gene ID: 8807. Restriction: For Research Use Only.
Catalog No. 50-150-1394 Supplier Biomatik Corporation Supplier No. EKL54560
The Human Interleukin 18 Receptor Accessory Protein (IL18RAP) ELISA Kit allows for quantitative determination in tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.051 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O95256. NCBI Gene ID: 8807. Restriction: For Research Use Only.