Learn More
Biomatik Corporation Human Interleukin 1 Alpha (IL1a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54569
Encompass_Preferred
The Human Interleukin 1 Alpha (IL1a) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 11.6 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P01583. NCBI Gene ID: 3552. Restriction: For Research Use Only.
Catalog No. 50-150-1398