Learn More
Biomatik Corporation Mouse Interleukin 1 Alpha (IL1a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54570
Encompass_Preferred
Mouse Interleukin 1 Alpha (IL1a) ELISA Kit for detecting IL1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P01582. Gene ID: 16175. Target Synonyms: IL1-A; IL-1alpha; IL1F1; Preinterleukin 1 Alpha; Hematopoietin-1; Pro-Interleukin-1-Alpha; Interleukin-1 Family Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1399