Learn More
Biomatik Corporation Mouse Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54597
Encompass_Preferred
Mouse Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit for detecting IL1RA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P25085. Gene ID: 16181. Target Synonyms: IL1RN; IL1-RA; ICIL1RA; IL1ra3; IL1F3; IRAP;IL-1RA; Interleukin-1 Family Member 3; IL1 inhibitor; Anakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1414