Learn More
Biomatik Corporation Human Interleukin 1 Zeta (IL1z) ELISA Kit, 96 tests
Catalog No. 501501416
Encompass_Preferred
The Human Interleukin 1 Zeta (IL1z) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 78.125-5000 pg/mL. Sensitivity: 24 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q9NZH6. NCBI Gene ID: 27178. Restriction: For Research Use Only.
Catalog No. 50-150-1416 Supplier Biomatik Corporation Supplier No. EKL54604
The Human Interleukin 1 Zeta (IL1z) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 78.125-5000 pg/mL. Sensitivity: 24 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q9NZH6. NCBI Gene ID: 27178. Restriction: For Research Use Only.