Learn More
Biomatik Corporation Human Interleukin 22 (IL22) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54623
Encompass_Preferred
The Human Interleukin 22 (IL22) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q9GZX6. NCBI Gene ID: 50616. Restriction: For Research Use Only.
Catalog No. 50-150-1424