Learn More
Biomatik Corporation Rat Interleukin 23 (IL23) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54630
Encompass_Preferred
Rat Interleukin 23 (IL23) ELISA Kit for detecting IL23 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q91Z84. Gene ID: 155140. Target Synonyms: IL23A; P19; IL23P19; SGRF; Interleukin-23 subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1428