Learn More
Biomatik Corporation Mouse Interleukin 6 (IL6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54698
Encompass_Preferred
Mouse Interleukin 6 (IL6) ELISA Kit for detecting IL6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2000pg/ml. Sensitivity: 14.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P08505. Gene ID: 16193. Target Synonyms: MGI2-A; MGI2A; HGF; BSF2; HSF; IFNB2; B-Cell Stimulatory Factor-2; Hybridoma/Plasmacytoma Growth Factor; Hepatocyte Stimulating Factor; Cytotoxic T-Cell Differentiation Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1457