Learn More
Biomatik Corporation Human Interleukin 7 Receptor (IL7R) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54710
Encompass_Preferred
The Human Interleukin 7 Receptor (IL7R) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.64 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P16871. NCBI Gene ID: 3575. Restriction: For Research Use Only.
Catalog No. 50-150-1462