Learn More
Biomatik Corporation Mouse Interleukin 9 (IL9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54730
Encompass_Preferred
Mouse Interleukin 9 (IL9) ELISA Kit for detecting IL9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P15247. Gene ID: 16198. Target Synonyms: HP40; P40; TCGFIII; TCGF3; T Cell Growth Factor III; P40 T-Cell And Mast Cell Growth Factor; P40 Cytokine; Homolog Of Mouse T Cell And Mast Cell Growth Factor 40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1471