Learn More
Biomatik Corporation Human Interferon Inducible T-Cell Alpha Chemoattractant (ITaC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54753
Encompass_Preferred
The Human Interferon Inducible T-Cell Alpha Chemoattractant (ITaC) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 10.8 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O14625. NCBI Gene ID: 6373. Restriction: For Research Use Only.
Catalog No. 50-150-1482