Learn More
Biomatik Corporation Mouse Klotho (KL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54762
Encompass_Preferred
Mouse Klotho (KL) ELISA Kit for detecting KL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: O35082. Gene ID: 16591. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1488