Learn More
Biomatik Corporation Human B-Lymphocyte Activation Antigen B7-2 (LAB7-2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54765
Encompass_Preferred
The Human B-Lymphocyte Activation Antigen B7-2 (LAB7-2) ELISA Kit allows for quantitative determination in tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P42081. NCBI Gene ID: 942. Restriction: For Research Use Only.
Catalog No. 50-150-149-1