Learn More
Biomatik Corporation Mouse Leptin (LEP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54769
Encompass_Preferred
Mouse Leptin (LEP) ELISA Kit for detecting LEP in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.103ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P41160. Gene ID: 16846. Target Synonyms: OB; OBS; Obesity Homolog; Obesity Factor; Obese Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-149-3