Learn More
Biomatik Corporation Human Lymphocyte Function Associated Antigen 3 (LFA3) ELISA Kit, 96 tests
Catalog No. 501501496
Encompass_Preferred
The Human Lymphocyte Function Associated Antigen 3 (LFA3) ELISA Kit allows for quantitative determination in serum, plasma, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 12.5-800 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P19256. NCBI Gene ID: 965. Restriction: For Research Use Only.
Catalog No. 50-150-149-6 Supplier Biomatik Corporation Supplier No. EKL54773
The Human Lymphocyte Function Associated Antigen 3 (LFA3) ELISA Kit allows for quantitative determination in serum, plasma, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 12.5-800 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P19256. NCBI Gene ID: 965. Restriction: For Research Use Only.