Learn More
Biomatik Corporation Rat Luteinizing Hormone (LH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54776
Encompass_Preferred
Rat Luteinizing Hormone (LH) ELISA Kit for detecting LH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.468-30ng/ml. Sensitivity: 0.14ng/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 4h, 30min. Target Synonyms: ICSH; Lutropin; Interstitial Cell Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-149-8