Learn More
Biomatik Corporation General Lipopolysaccharides (LPS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54778
Encompass_Preferred
The General Lipopolysaccharides (LPS) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: All species. Detection Range: 3.12-200 ng/mL. Sensitivity: 1.24 ng/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-149-9