Learn More
Biomatik Corporation Human Low Density Lipoprotein Receptor Related Protein 1 (LRP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54779
Encompass_Preferred
The Human Low Density Lipoprotein Receptor Related Protein 1 (LRP1) ELISA Kit allows for quantitative determination in serum, plasma or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q07954. NCBI Gene ID: 4035. Restriction: For Research Use Only.
Catalog No. 50-150-1500