Learn More
Biomatik Corporation Human Latent Transforming Growth Factor Beta Binding Protein 2 (LTBP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54785
Encompass_Preferred
The Human Latent Transforming Growth Factor Beta Binding Protein 2 (LTBP2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q14767. NCBI Gene ID: 4053. Restriction: For Research Use Only.
Catalog No. 50-150-1503