Learn More
Biomatik Corporation Human Colony Stimulating Factor 1, Macrophage (MCSF) ELISA Kit, 96 tests
Catalog No. 501501507
Encompass_Preferred
The Human Colony Stimulating Factor 1, Macrophage (MCSF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P09603. NCBI Gene ID: 1435. Restriction: For Research Use Only.
Catalog No. 50-150-1507 Supplier Biomatik Corporation Supplier No. EKL54792
The Human Colony Stimulating Factor 1, Macrophage (MCSF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P09603. NCBI Gene ID: 1435. Restriction: For Research Use Only.