Learn More
Biomatik Corporation Human Monokine Induced By Interferon Gamma (MIg) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54798
Encompass_Preferred
The Human Monokine Induced By Interferon Gamma (MIg) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. Restriction: For Research Use Only.
Catalog No. 50-150-1513