Learn More
Biomatik Corporation Rat Monokine Induced By Interferon Gamma (MIg) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54800
Encompass_Preferred
Rat Monokine Induced By Interferon Gamma (MIg) ELISA Kit for detecting MIg in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q8K4B1. Gene ID: 246759. Target Synonyms: CXCL9; CMK; Humig; MI-G; SCYB9; Crg-10; Chemokine (C-X-C motif) ligand 9; Small-inducible cytokine B9; Gamma-interferon-induced monokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1514