Learn More
Biomatik Corporation Rat Myeloperoxidase (MPO) ELISA Kit, 96 tests
Catalog No. 501501519
Encompass_Preferred
Rat Myeloperoxidase (MPO) ELISA Kit for detecting MPO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 13.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: D3ZYH8. Gene ID: 303413. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1519 Supplier Biomatik Corporation Supplier No. EKL54808
Rat Myeloperoxidase (MPO) ELISA Kit for detecting MPO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 13.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: D3ZYH8. Gene ID: 303413. Restrictions: For Research Use Only. Not for use in diagnostic procedures.