Learn More
Biomatik Corporation Human NADH Dehydrogenase 1 (ND1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54820
Encompass_Preferred
The Human NADH Dehydrogenase 1 (ND1) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.068 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. Restriction: For Research Use Only.
Catalog No. 50-150-1523