Learn More
Biomatik Corporation Mouse Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54823
Encompass_Preferred
Mouse Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit for detecting NGAL in serum, plasma, tissue homogenates, urine, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.285ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P11672. Gene ID: 16819. Target Synonyms: LCN2; p25; Lipocalin 2; Oncogene 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; Siderocalin LCN2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1525