Learn More
Biomatik Corporation Rat Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit, 96 tests
Catalog No. 501501526
Encompass_Preferred
Rat Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit for detecting NGAL in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.1-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P30152. Gene ID: 170496. Target Synonyms: LCN2; p25; Lipocalin 2; Oncogene 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; Siderocalin LCN2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1526 Supplier Biomatik Corporation Supplier No. EKL54824
Rat Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit for detecting NGAL in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.1-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P30152. Gene ID: 170496. Target Synonyms: LCN2; p25; Lipocalin 2; Oncogene 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; Siderocalin LCN2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.