Learn More
Biomatik Corporation Mouse Nerve Growth Factor (NGF) ELISA Kit, 96 tests
Catalog No. 501501528
Encompass_Preferred
Mouse Nerve Growth Factor (NGF) ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P01139. Gene ID: 18049. Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1528 Supplier Biomatik Corporation Supplier No. EKL54829
Mouse Nerve Growth Factor (NGF) ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P01139. Gene ID: 18049. Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.