Learn More
Biomatik Corporation Rat Osteopontin (OPN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54853
Encompass_Preferred
Rat Osteopontin (OPN) ELISA Kit for detecting OPN in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.32ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P08721. Gene ID: 25353. Target Synonyms: SPP1; BNSP; BSPI; ETA1; Secreted Phosphoprotein 1; Bone Sialoprotein I; Early T-Lymphocyte Activation 1; Nephropontin; Urinary stone protein; Uropontin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1538