Learn More
Biomatik Corporation General Platelet Activating Factor (PAF) ELISA Kit, 96 tests
Catalog No. 501501539
Encompass_Preferred
The General Platelet Activating Factor (PAF) ELISA Kit allows for quantitative determination in serum, plasma or other biological fluids. Species Reactivity: All species. Detection Range: 0.468-30 ng/mL. Sensitivity: 0.224 ng/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-1539 Supplier Biomatik Corporation Supplier No. EKL54854
The General Platelet Activating Factor (PAF) ELISA Kit allows for quantitative determination in serum, plasma or other biological fluids. Species Reactivity: All species. Detection Range: 0.468-30 ng/mL. Sensitivity: 0.224 ng/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.