Learn More
Biomatik Corporation Mouse Plasminogen Activator Inhibitor 1 (PAI1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54855
Encompass_Preferred
Mouse Plasminogen Activator Inhibitor 1 (PAI1) ELISA Kit for detecting PAI1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 23.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P22777. Gene ID: 18787. Target Synonyms: SERPINE1; PLANH1; Serpin Peptidase Inhibitor Clade E Member 1; Nexin; Plasminogen Activator Inhibitor Type 1 Serpin E1; Endothelial plasminogen activator inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1540