Learn More
Biomatik Corporation Mouse Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit, 96 tests
Catalog No. 501501543
Encompass_Preferred
Mouse Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit for detecting PDGFA in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P20033. Gene ID: 18590. Target Synonyms: PDGF-A; PDGF1; Platelet Derived Growth Factor Alpha Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1543 Supplier Biomatik Corporation Supplier No. EKL54863
Mouse Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit for detecting PDGFA in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P20033. Gene ID: 18590. Target Synonyms: PDGF-A; PDGF1; Platelet Derived Growth Factor Alpha Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.