Learn More
Biomatik Corporation Human Platelet Derived Growth Factor Subunit B (PDGFB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54867
Encompass_Preferred
The Human Platelet Derived Growth Factor Subunit B (PDGFB) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 11.3 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P01127. NCBI Gene ID: 5155. Restriction: For Research Use Only.
Catalog No. 50-150-1546