Learn More
Biomatik Corporation Rat Lipase, Pancreatic (PL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54885
Encompass_Preferred
Rat Lipase, Pancreatic (PL) ELISA Kit for detecting PL in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 37pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P27657. Gene ID: 25702. Target Synonyms: PNLIP; PTL; Pancreatic Lipase; Pancreatic triacylglycerol lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1553