Learn More
Biomatik Corporation Mouse Placenta Growth Factor (PLGF) ELISA Kit, 96 tests
Catalog No. 501501555
Encompass_Preferred
Mouse Placenta Growth Factor (PLGF) ELISA Kit for detecting PLGF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P49764. Gene ID: 18654. Target Synonyms: PlGF2; PGF; PGFL; Placental Growth Factor-Like; Vascular Endothelial Growth Factor-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1555 Supplier Biomatik Corporation Supplier No. EKL54888
Mouse Placenta Growth Factor (PLGF) ELISA Kit for detecting PLGF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P49764. Gene ID: 18654. Target Synonyms: PlGF2; PGF; PGFL; Placental Growth Factor-Like; Vascular Endothelial Growth Factor-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.