Learn More
Biomatik Corporation Rat Regenerating Islet Derived Protein 1 Alpha (REG1a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54907
Encompass_Preferred
Rat Regenerating Islet Derived Protein 1 Alpha (REG1a) ELISA Kit for detecting REG1a in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P10758. Gene ID: 24714. Target Synonyms: REG; PSP; ICRF; PSPS1; PTP; Islet cells regeneration factor; Islet of Langerhans regenerating protein; Lithostathine 1 Alpha; Pancreatic Stone Protein; Pancreatic Thread Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1559