Learn More
Biomatik Corporation Human S100 Calcium Binding Protein A8 (S100A8) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKL54918
Human S100 Calcium Binding Protein A8 (S100A8) ELISA Kit for detecting S100A8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 26pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P05109. Gene ID: 6279. Target Synonyms: MRP8; MIF; 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; NIF; P8; Calgranulin A; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.