Learn More
Biomatik Corporation Mouse S100 Calcium Binding Protein B (S100B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54920
Encompass_Preferred
Mouse S100 Calcium Binding Protein B (S100B) ELISA Kit for detecting S100B in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P50114. Gene ID: 20203. Target Synonyms: S100-B; NEF; S100Beta; S-100 protein subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1565