Learn More
Biomatik Corporation Rat S100 Calcium Binding Protein B (S100B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54921
Encompass_Preferred
Rat S100 Calcium Binding Protein B (S100B) ELISA Kit for detecting S100B in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P04631. Gene ID: 25742. Target Synonyms: S100-B; NEF; S100Beta; S-100 protein subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1566