Learn More
Biomatik Corporation Human Semaphorin 3F (SEMA3F) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54928
Encompass_Preferred
Human Semaphorin 3F (SEMA3F) ELISA Kit for detecting SEMA3F in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.875-3000pg/ml. Sensitivity: 19.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q13275. Gene ID: 6405. Target Synonyms: SEMA-IV; SEMA4; SEMAK; Sema Domain; Immunoglobulin Domain(Ig); Transmembrane Domain(TM)and Short Cytoplasmic Domain 3F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1570