Learn More
Biomatik Corporation Human Synaptopodin (SYNPO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54946
Encompass_Preferred
Human Synaptopodin (SYNPO) ELISA Kit for detecting SYNPO in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q8N3V7. Gene ID: 11346. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1577