Learn More
Biomatik Corporation Mouse Transforming Growth Factor Alpha (TGFa) ELISA Kit, 96 tests
Catalog No. 501501580
Encompass_Preferred
Mouse Transforming Growth Factor Alpha (TGFa) ELISA Kit for detecting TGFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P48030. Gene ID: 21802. Target Synonyms: ETGF; TGF-A; Protransforming Growth Factor alpha; EGF-Like TGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1580 Supplier Biomatik Corporation Supplier No. EKL54958
Mouse Transforming Growth Factor Alpha (TGFa) ELISA Kit for detecting TGFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P48030. Gene ID: 21802. Target Synonyms: ETGF; TGF-A; Protransforming Growth Factor alpha; EGF-Like TGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.