Learn More
Biomatik Corporation Human Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54974
Encompass_Preferred
Human Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit for detecting TGFb2 in serum, plasma, tissue homogenates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 8.64pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P61812. Gene ID: 7042. Target Synonyms: TGF-B2; TGF-Beta2; G-TSF; LAP; Cetermin; Polyergin; Latency-associated peptide; BSC-1 cell growth inhibitor; Glioblastoma-derived T-cell suppressor factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1584