Learn More
Biomatik Corporation Mouse Thrombospondin 1 (THBS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54993
Encompass_Preferred
Mouse Thrombospondin 1 (THBS1) ELISA Kit for detecting THBS1 in plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 6.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P35441. Gene ID: 21825. Target Synonyms: TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1591