Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Alpha (TNFa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55003
Encompass_Preferred
Mouse Tumor Necrosis Factor Alpha (TNFa) ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P06804. Gene ID: 21926. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2; TNF; Tumor Necrosis Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1597