Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Alpha Induced Protein 6 (TNFaIP6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55010
Encompass_Preferred
Mouse Tumor Necrosis Factor Alpha Induced Protein 6 (TNFaIP6) ELISA Kit for detecting TNFaIP6 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: O08859. Gene ID: 21930. Target Synonyms: TSG6; TNF-Stimulated Gene 6; Hyaluronate-binding protein; Tumor necrosis factor alpha-induced protein 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1600