Learn More
Biomatik Corporation Human Tumor Necrosis Factor Ligand Superfamily, Member 9 (TNFSF9) ELISA Kit, 96 tests
Catalog No. 501501614
Encompass_Preferred
Human Tumor Necrosis Factor Ligand Superfamily, Member 9 (TNFSF9) ELISA Kit for detecting TNFSF9 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.35pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P41273. Gene ID: 8744. Target Synonyms: 4-1BB-L; Receptor 4-1BB Ligand; Homolog Of Mouse 4-1BB-L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1614 Supplier Biomatik Corporation Supplier No. EKL55037
Human Tumor Necrosis Factor Ligand Superfamily, Member 9 (TNFSF9) ELISA Kit for detecting TNFSF9 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.35pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P41273. Gene ID: 8744. Target Synonyms: 4-1BB-L; Receptor 4-1BB Ligand; Homolog Of Mouse 4-1BB-L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.